Hexachloro-1,3-butadiene 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-484
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Cyclohexane
- Description:Hexachloro-1,3-butadiene 100 µg/mL in Cyclohexane
- Size:1 mL
- Cat. No.:77696-484