EPA Method 8260 VOC Gases Mixture 2000 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-535
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- Description:EPA Method 8260 VOC Gases Mixture 2000 µg/mL in Methanol
- Size:1 mL
- Cat. No.:77696-535