$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Melting Point Standards
Benzil - Melting Point 94.90 ºC, LGC, LGC Standards
Benzil - Melting Point 94.90 ºC, LGC, LGC Standards
Catalog # 77691-934
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Benzil - Melting Point 94.90 ºC, LGC, LGC Standards
Catalog # 77691-934
Supplier:
LGC Standards
Supplier Number:
ERM-FC027
New Product
CAS Number:
Specifications
Description:
Benzil - Melting Point 94.90 ºC
Size:
0.25 g
Cat. No.:
77691-934
Specifications
About this item
Product Details
Product Details