Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Melting Point Standards
4-Nitrotoluene - Melting Point 51.66 ºC, LGC, LGC Standards
4-Nitrotoluene - Melting Point 51.66 ºC, LGC, LGC Standards
4-Nitrotoluene - Melting Point 51.66 ºC, LGC, LGC Standards
Catalog # 77691-935
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
4-Nitrotoluene - Melting Point 51.66 ºC, LGC, LGC Standards
Catalog # 77691-935
Supplier:
LGC Standards
Supplier Number:
ERM-FC029
New Product
CAS Number:
Specifications
Description:
4-Nitrotoluene - Melting Point 51.66 ºC
Size:
0.25 g
Cat. No.:
77691-935
Specifications
About this item
Product Details
Product Details