$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Food Standards
Ethanol 300mg/100mL in Water, LoGiCal, LGC Standards
Ethanol 300mg/100mL in Water, LoGiCal, LGC Standards
77697-029
:
LGC Standards
New Product
:
Ethanol 300mg/100mL in Water, LoGiCal, LGC Standards
77697-029
:
LGC Standards
:
LGCETH-300
New Product
:
Matrix:
Water
Description:
Ethanol 300mg/100mL in Water
Size:
1 mL
Cat. No.:
77697-029