$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Food Standards
Ethanol 500mg/100mL in Water, LoGiCal, LGC Standards
Ethanol 500mg/100mL in Water, LoGiCal, LGC Standards
Ethanol 500mg/100mL in Water, LoGiCal, LGC Standards
Catalog # 77697-031
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Ethanol 500mg/100mL in Water, LoGiCal, LGC Standards
Catalog # 77697-031
Supplier:
LGC Standards
Supplier Number:
LGCETH-500
New Product
CAS Number:
Specifications
Matrix:
Water
Description:
Ethanol 500mg/100mL in Water
Size:
1 mL
Cat. No.:
77697-031
Specifications
About this item
Product Details
Product Details