Spectrometric Oil Standard D12-010: Ag, Al, Cr, Cu, Fe, Mg, Na, Ni, Pb, Si, Sn, Ti @ 10 µg/g in Aviation Reference Oil, VHG, LGC Standards
Catalog # 77697-449
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Aviation Oil
- Description:Spectrometric Oil Standard D12-010: Ag, Al, Cr, Cu, Fe, Mg, Na, Ni, Pb, Si, Sn, Ti @ 10 µg/g in Aviation Reference Oil
- Size:200 g
- Cat. No.:77697-449