Spectrometric Oil Standard D3-100: B, Mo, Zn @ 100 µg/g in Aviation Reference Oil, VHG, LGC Standards
Catalog # 77697-450
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Aviation Oil
- Description:Spectrometric Oil Standard D3-100: B, Mo, Zn @ 100 µg/g in Aviation Reference Oil
- Size:240 mL
- Cat. No.:77697-450