Base Number (BN) Standard: 6 mg KOH/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77693-049
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Hydrocarbon Oil
- Description:Base Number (BN) Standard: 6 mg KOH/g in Hydrocarbon Oil
- Size:50 g
- Cat. No.:77693-049