Crackle Test Reference Standard: 0 wt% H2O in 10W30 Motor Oil, VHG, LGC Standards
Catalog # 77693-078
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:10W30 Motor Oil
- Description:Crackle Test Reference Standard: 0 wt% H2O in 10W30 Motor Oil
- Size:100 mL
- Cat. No.:77693-078