ContactUsLinkComponent
Spectrometric Oil Reference Standard D19-300: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 300 µg/g in Aviation Reference Oil, VHG, LGC Standards
Spectrometric Oil Reference Standard D19-300: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 300 µg/g in Aviation Reference Oil, VHG, LGC Standards
  77697-505
New Product
 :  
undefined
Spectrometric Oil Reference Standard D19-300: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 300 µg/g in Aviation Reference Oil, VHG, LGC Standards
  77697-505
 :  VHG-D19-300-100G
New Product
 :  
  • Matrix:
    Aviation Oil
  • Description:
    Spectrometric Oil Reference Standard D19-300: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 300 µg/g in Aviation Reference Oil
  • Size:
    100 g
  • Cat. No.:
    77697-505