Order Entry
Puerto Rico
ContactUsLinkComponent
Spectrometric Oil Reference Standard D19-80: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 80 µg/g in Aviation Reference Oil, VHG, LGC Standards
Spectrometric Oil Reference Standard D19-80: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 80 µg/g in Aviation Reference Oil, VHG, LGC Standards
Catalog # 77697-511
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Spectrometric Oil Reference Standard D19-80: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 80 µg/g in Aviation Reference Oil, VHG, LGC Standards
Catalog # 77697-511
Supplier:  LGC Standards
Supplier Number:  VHG-D19-80-100G
New Product
CAS Number:  

Specifications

  • Matrix:
    Aviation Oil
  • Description:
    Spectrometric Oil Reference Standard D19-80: Ag, Al, B, Ba, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Si, Sn, Ti, V, Zn @ 80 µg/g in Aviation Reference Oil
  • Size:
    100 g
  • Cat. No.:
    77697-511

Specifications

About this item