10% (w/w) Devolatilized Diesel Fuel in 75 cSt Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77693-083
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Hydrocarbon Oil
- Description:10% (w/w) Devolatilized Diesel Fuel in 75 cSt Hydrocarbon Oil
- Size:100 mL
- Cat. No.:77693-083