Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
Red Dye #26 QC Check 10 mg/L, VHG, LGC Standards
Red Dye #26 QC Check 10 mg/L, VHG, LGC Standards
77697-514
:
LGC Standards
New Product
:
Red Dye #26 QC Check 10 mg/L, VHG, LGC Standards
77697-514
:
LGC Standards
:
VHG-DSLRED26-QC-100
New Product
:
Matrix:
Kerosene
Description:
Red Dye #26 QC Check 10 mg/L
Size:
100 mL
Cat. No.:
77697-514