5% (w/w) Devolatilized Gasoline in 75 cSt Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77693-098
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:75 cSt Hydrocarbon Oil
- Description:5% (w/w) Devolatilized Gasoline in 75 cSt Hydrocarbon Oil
- Size:100 mL
- Cat. No.:77693-098