Multi-Element Coolant Standard: P @ 2500; B, K, Na @ 1000; Mo, Si @ 500; Al, Ca, Cu, Fe, Mg, Pb, Zn @ 50 μg/g in dil. HNO3, tr. HF, VHG, LGC Standards
Catalog # 77697-523
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Dilute HNO3
- Description:Multi-Element Coolant Standard: P @ 2500; B, K, Na @ 1000; Mo, Si @ 500; Al, Ca, Cu, Fe, Mg, Pb, Zn @ 50 μg/g in dil. HNO3, tr. HF
- Size:500 mL
- Cat. No.:77697-523