Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
NH3 as N: 1000µg/mL in H20, VHG, LGC Standards
NH3 as N: 1000µg/mL in H20, VHG, LGC Standards
NH3 as N: 1000µg/mL in H20, VHG, LGC Standards
Catalog # 77697-575
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
NH3 as N: 1000µg/mL in H20, VHG, LGC Standards
Catalog # 77697-575
Supplier:
LGC Standards
Supplier Number:
VHG-INH3N-100
New Product
CAS Number:
Specifications
Matrix:
H2O
Description:
NH3 as N: 1000µg/mL in H20
Size:
100 mL
Cat. No.:
77697-575
Specifications
About this item
Product Details
Product Details