Polysulfide Oil Standard: S @ 25 µg/g in 13 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77697-819
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:13 cSt Mineral Oil
- Description:Polysulfide Oil Standard: S @ 25 µg/g in 13 cSt Mineral Oil
- Size:1 L
- Cat. No.:77697-819