Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
9-12 wt% Soot in Diesel Engine Oil, VHG, LGC Standards
9-12 wt% Soot in Diesel Engine Oil, VHG, LGC Standards
9-12 wt% Soot in Diesel Engine Oil, VHG, LGC Standards
Catalog # 77693-271
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
9-12 wt% Soot in Diesel Engine Oil, VHG, LGC Standards
Catalog # 77693-271
Supplier:
LGC Standards
Supplier Number:
VHG-SOOT-E-50
New Product
CAS Number:
Specifications
Matrix:
15W40 motor oil
Description:
9-12 wt% Soot in Diesel Engine Oil
Size:
50 mL
Cat. No.:
77693-271
Specifications
About this item
Product Details
Product Details