Mercaptan Sulfur Set for UOP 163 and ASTM D3227: S @ 10 µg/g in 80% Isooctane/20% Toluene, VHG, LGC Standards
Catalog # 77697-917
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Isooctane:Toluene
- Description:Mercaptan Sulfur Set for UOP 163 and ASTM D3227: S @ 10 µg/g in 80% Isooctane/20% Toluene
- Size:6 x 20 ml
- Cat. No.:77697-917