Order Entry
Puerto Rico
ContactUsLinkComponent
Viscosity Standard: 100 cSt at 40°C, and 16.8 cSt at 100°C, VHG, LGC Standards
Viscosity Standard: 100 cSt at 40°C, and 16.8 cSt at 100°C, VHG, LGC Standards
Catalog # 77695-703
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Viscosity Standard: 100 cSt at 40°C, and 16.8 cSt at 100°C, VHG, LGC Standards
Catalog # 77695-703
Supplier:  LGC Standards
Supplier Number:  VHG-VISC100A-500
New Product
CAS Number:  

Specifications

  • Description:
    Viscosity Standard: 100 cSt at 40°C, and 16.8 cSt at 100°C
  • Size:
    500 ml
  • Cat. No.:
    77695-703

Specifications

About this item