Anti-CDK5RAP2 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: CDK5RAP2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77724-046
- 77724-045
- Antigen Symbol:
- CDK5RAP2
- CDK5RAP2
- Antigen Synonyms:
- CDK5RAP2|CDK5 regulatory subunit-associated protein 2|CDK5 regulatory subunit associated protein 2|C48|CEP215|MCPH3|Centrosome-associated protein 215|CDK5 activator-binding protein C48|centrosomal protein 215 kDa|CK5P2_HUMAN
- CDK5RAP2|CDK5 regulatory subunit-associated protein 2|CDK5 regulatory subunit associated protein 2|C48|CEP215|MCPH3|Centrosome-associated protein 215|CDK5 activator-binding protein C48|centrosomal protein 215 kDa|CK5P2_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96SN8
- Q96SN8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human CDK5 regulatory subunit-associated protein 2 protein (996-1111AA)
- Recombinant Human CDK5 regulatory subunit-associated protein 2 protein (996-1111AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice