Anti-CHMP6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200
Type:
Antigen: CHMP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat
Specifications
- Catalog No:
- 77723-815
- 77690-363
- Antigen Symbol:
- CHMP6
- CHMP6
- Antigen Synonyms:
- RP24 225K18.4|Charged multivesicular body protein 6|hVps 20|OTTMUSP00000004363|Vps 20|Vps20|Vacuolar protein sorting associated protein 20|hVps20|Vacuolar protein sorting-associated protein 20|Chromatin modifying protein 6|RGD1565325|FLJ11749|Chromatin-modifying protein 6|novel SNF7 protein|CHMP6_HUMAN|chmp6
- RP24 225K18.4|Charged multivesicular body protein 6|hVps 20|OTTMUSP00000004363|Vps 20|Vps20|Vacuolar protein sorting associated protein 20|hVps20|Vacuolar protein sorting-associated protein 20|Chromatin modifying protein 6|RGD1565325|FLJ11749|Chromatin-modifying protein 6|novel SNF7 protein|CHMP6_HUMAN|chmp6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96FZ7
- Q96FZ7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Charged multivesicular body protein 6 protein (2-201AA)
- Recombinant Human Charged multivesicular body protein 6 protein (2-201AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice