Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-RIT1 Rabbit Polyclonal Antibody (HRP)
Anti-RIT1 Rabbit Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-RIT1 Rabbit Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77723-633
  • 77723-632
  • Antigen Symbol:
  • RIT1
  • RIT1
  • Antigen Synonyms:
  • RIT|Ric-like protein without CAAX motif 1|NS8|RIBB|Ras-like protein expressed in many tissues|GTP-binding protein Rit1|Ras like without CAAX 1|GTP binding protein Roc1|Ric like expressed in many tissues|RIT1|RIT1_HUMAN|Ras-like without CAAX protein 1|ROC1|Ras like protein expressed in many tissues
  • RIT|Ric-like protein without CAAX motif 1|NS8|RIBB|Ras-like protein expressed in many tissues|GTP-binding protein Rit1|Ras like without CAAX 1|GTP binding protein Roc1|Ric like expressed in many tissues|RIT1|RIT1_HUMAN|Ras-like without CAAX protein 1|ROC1|Ras like protein expressed in many tissues
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q92963
  • Q92963
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human GTP-binding protein Rit1 protein (1-219AA)
  • Recombinant Human GTP-binding protein Rit1 protein (1-219AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue Ice
  • Shipped on Blue Ice

Specifications