Anti-LMO3 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LMO3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-326
- 77723-325
- Antigen Symbol:
- LMO3
- LMO3
- Antigen Synonyms:
- LIM domain only protein 3|RHOM 3|lmo3|RBTNL 2|MGC26081|LMO 3|DAT1|RBTN3|RHOM3|RBTN 3|DAT 1|Neuronal specific transcription factor DAT1|Rhombotin 3|LMO3_HUMAN|LMO-3|Rhombotin like 2|Rhombotin3|RBTNL2|Rhombotin-3|LIM domain only 3 (rhombotin like 2)|Neuronal-specific transcription factor DAT1
- LIM domain only protein 3|RHOM 3|lmo3|RBTNL 2|MGC26081|LMO 3|DAT1|RBTN3|RHOM3|RBTN 3|DAT 1|Neuronal specific transcription factor DAT1|Rhombotin 3|LMO3_HUMAN|LMO-3|Rhombotin like 2|Rhombotin3|RBTNL2|Rhombotin-3|LIM domain only 3 (rhombotin like 2)|Neuronal-specific transcription factor DAT1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TAP4
- Q8TAP4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human LIM domain only protein 3 protein (1-145AA)
- Recombinant Human LIM domain only protein 3 protein (1-145AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice