Anti-HSD17B11 Rabbit Polyclonal Antibody (Biotin)
New Product
Type:
Antigen: HSD17B11
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77723-166
- 77723-165
- Antigen Symbol:
- HSD17B11
- HSD17B11
- Antigen Synonyms:
- Estradiol 17-beta-dehydrogenase 11|17betaHSDXI|Hsd17b11|Hydroxysteroid (17 beta) dehydrogenase 11|17-beta-hydroxysteroid dehydrogenase XI|17 beta hydroxysteroid dehydrogenase XI|PAN1B|17 beta hydroxysteroid dehydrogenase 11|17betaHSD11|Retinal short-chain dehydrogenase/reductase 2|Estradiol 17 beta dehydrogenase 11|17-beta-HSD XI|17 beta hydroxysteroid dehydrogenase type XI|17bHSD11|17-beta-hydroxysteroid dehydrogenase 11|CTCL associated antigen HD CL 03|RETSDR2|Cutaneous T-cell lymphoma-associated antigen HD-CL-03|SDR16C2|SDR2|Short chain dehydrogenase/reductase family 16C member 2|Cutaneous T cell lymphoma associated antigen HD CL 03|T cell lymphoma associated antigen HD CL 03|17 beta HSD XI|17-beta-HSD 11|17 BETA HSDXI |DHRS8|CTCL-associated antigen HD-CL-03|17 beta HSD 11|DHB11_HUMAN|Retinal short chain dehydrogenase/reductase 2|CTCL tumor antigen HD CL 03 |Dehydrogenase/reductase SDR family member 8|17 BETA HSD11
- Estradiol 17-beta-dehydrogenase 11|17betaHSDXI|Hsd17b11|Hydroxysteroid (17 beta) dehydrogenase 11|17-beta-hydroxysteroid dehydrogenase XI|17 beta hydroxysteroid dehydrogenase XI|PAN1B|17 beta hydroxysteroid dehydrogenase 11|17betaHSD11|Retinal short-chain dehydrogenase/reductase 2|Estradiol 17 beta dehydrogenase 11|17-beta-HSD XI|17 beta hydroxysteroid dehydrogenase type XI|17bHSD11|17-beta-hydroxysteroid dehydrogenase 11|CTCL associated antigen HD CL 03|RETSDR2|Cutaneous T-cell lymphoma-associated antigen HD-CL-03|SDR16C2|SDR2|Short chain dehydrogenase/reductase family 16C member 2|Cutaneous T cell lymphoma associated antigen HD CL 03|T cell lymphoma associated antigen HD CL 03|17 beta HSD XI|17-beta-HSD 11|17 BETA HSDXI |DHRS8|CTCL-associated antigen HD-CL-03|17 beta HSD 11|DHB11_HUMAN|Retinal short chain dehydrogenase/reductase 2|CTCL tumor antigen HD CL 03 |Dehydrogenase/reductase SDR family member 8|17 BETA HSD11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NBQ5
- Q8NBQ5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Estradiol 17-beta-dehydrogenase 11 protein (20-300AA)
- Recombinant Human Estradiol 17-beta-dehydrogenase 11 protein (20-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice