Anti-HSD17B11 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HSD17B11
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77723-161
- 77723-162
- Antigen Symbol:
- HSD17B11
- HSD17B11
- Antigen Synonyms:
- Estradiol 17-beta-dehydrogenase 11|17betaHSDXI|Hsd17b11|Hydroxysteroid (17 beta) dehydrogenase 11|17-beta-hydroxysteroid dehydrogenase XI|17 beta hydroxysteroid dehydrogenase XI|PAN1B|17 beta hydroxysteroid dehydrogenase 11|17betaHSD11|Retinal short-chain dehydrogenase/reductase 2|Estradiol 17 beta dehydrogenase 11|17-beta-HSD XI|17 beta hydroxysteroid dehydrogenase type XI|17bHSD11|17-beta-hydroxysteroid dehydrogenase 11|CTCL associated antigen HD CL 03|RETSDR2|Cutaneous T-cell lymphoma-associated antigen HD-CL-03|SDR16C2|SDR2|Short chain dehydrogenase/reductase family 16C member 2|Cutaneous T cell lymphoma associated antigen HD CL 03|T cell lymphoma associated antigen HD CL 03|17 beta HSD XI|17-beta-HSD 11|17 BETA HSDXI |DHRS8|CTCL-associated antigen HD-CL-03|17 beta HSD 11|DHB11_HUMAN|Retinal short chain dehydrogenase/reductase 2|CTCL tumor antigen HD CL 03 |Dehydrogenase/reductase SDR family member 8|17 BETA HSD11
- Estradiol 17-beta-dehydrogenase 11|17betaHSDXI|Hsd17b11|Hydroxysteroid (17 beta) dehydrogenase 11|17-beta-hydroxysteroid dehydrogenase XI|17 beta hydroxysteroid dehydrogenase XI|PAN1B|17 beta hydroxysteroid dehydrogenase 11|17betaHSD11|Retinal short-chain dehydrogenase/reductase 2|Estradiol 17 beta dehydrogenase 11|17-beta-HSD XI|17 beta hydroxysteroid dehydrogenase type XI|17bHSD11|17-beta-hydroxysteroid dehydrogenase 11|CTCL associated antigen HD CL 03|RETSDR2|Cutaneous T-cell lymphoma-associated antigen HD-CL-03|SDR16C2|SDR2|Short chain dehydrogenase/reductase family 16C member 2|Cutaneous T cell lymphoma associated antigen HD CL 03|T cell lymphoma associated antigen HD CL 03|17 beta HSD XI|17-beta-HSD 11|17 BETA HSDXI |DHRS8|CTCL-associated antigen HD-CL-03|17 beta HSD 11|DHB11_HUMAN|Retinal short chain dehydrogenase/reductase 2|CTCL tumor antigen HD CL 03 |Dehydrogenase/reductase SDR family member 8|17 BETA HSD11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NBQ5
- Q8NBQ5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Estradiol 17-beta-dehydrogenase 11 protein (20-300AA)
- Recombinant Human Estradiol 17-beta-dehydrogenase 11 protein (20-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice