Anti-ABHD14B Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: ABHD14B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
- Catalog No:
- 77722-698
- 77722-699
- Antigen Symbol:
- ABHD14B
- ABHD14B
- Antigen Synonyms:
- CCG1 interacting factor B|CCG1-interacting factor B|MGC15429|ABHD14B|OTTHUMP00000212469|CIB|Abhydrolase domain-containing protein 14B|Cell cycle gene 1 interacting factor B|Abhydrolase domain containing protein 14B|ABHEB_HUMAN|Abhydrolase domain containing 14B
- CCG1 interacting factor B|CCG1-interacting factor B|MGC15429|ABHD14B|OTTHUMP00000212469|CIB|Abhydrolase domain-containing protein 14B|Cell cycle gene 1 interacting factor B|Abhydrolase domain containing protein 14B|ABHEB_HUMAN|Abhydrolase domain containing 14B
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96IU4
- Q96IU4
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein ABHD14B protein (2-210AA)
- Recombinant Human Protein ABHD14B protein (2-210AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice