Anti-ACY3 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: ACY3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77722-692
- 77722-693
- Antigen Symbol:
- ACY3
- ACY3
- Antigen Synonyms:
- Aminoacylase III|EC 3.5.1.15|acy3|EC 3.5.1.-|AA3|AW107362|HCBP1|ACY-3|Hepatitis C virus core-binding protein 1|ACY3_HUMAN|Aminoacylase-3|Aspartoacylase (aminoacylase) 3|Aspartoacylase-2|Hepatitis C virus core binding protein 1|Acylase III|ASPA2|Aminoacylase 3|Aspartoacylase 2
- Aminoacylase III|EC 3.5.1.15|acy3|EC 3.5.1.-|AA3|AW107362|HCBP1|ACY-3|Hepatitis C virus core-binding protein 1|ACY3_HUMAN|Aminoacylase-3|Aspartoacylase (aminoacylase) 3|Aspartoacylase-2|Hepatitis C virus core binding protein 1|Acylase III|ASPA2|Aminoacylase 3|Aspartoacylase 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96HD9
- Q96HD9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming) protein (5-319AA)
- Recombinant Human N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming) protein (5-319AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice