Anti-UBE2W Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBE2W
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-560
- 77722-561
- Antigen Symbol:
- UBE2W
- UBE2W
- Antigen Synonyms:
- UBC 16|ube2w|hUBC 16|FLJ11011|Ubiquitin protein ligase W|Ubiquitin conjugating enzyme E2W|hUBC16|Probable ubiquitin-conjugating enzyme E2 W|Ubiquitin conjugating enzyme E2 W|UBE2W_HUMAN|UBC16|Ubiquitin-protein ligase W|Probable ubiquitin conjugating enzyme E2 W|Ubiquitin carrier protein W|Ubiquitin conjugating enzyme 16|UBE 2W|UBC-16
- UBC 16|ube2w|hUBC 16|FLJ11011|Ubiquitin protein ligase W|Ubiquitin conjugating enzyme E2W|hUBC16|Probable ubiquitin-conjugating enzyme E2 W|Ubiquitin conjugating enzyme E2 W|UBE2W_HUMAN|UBC16|Ubiquitin-protein ligase W|Probable ubiquitin conjugating enzyme E2 W|Ubiquitin carrier protein W|Ubiquitin conjugating enzyme 16|UBE 2W|UBC-16
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96B02
- Q96B02
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin-conjugating enzyme E2 W protein (1-151AA)
- Recombinant Human Ubiquitin-conjugating enzyme E2 W protein (1-151AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice