Anti-BCL2L1 (phospho–Ser62) Rabbit Polyclonal Antibody
77716-030
New Product
- Antigen Symbol:BCL2L1 (phospho-Ser62)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Form:liquid
- Gene ID:Q07817
- Antigen Synonyms:Apoptosis regulator BclX|BclXL|Bcl 2 like 1 protein|Bcl X|B2CL1_HUMAN|BCLX|Bcl2 related gene|Protein phosphatase 1 regulatory subunit 52|Apoptosis regulator Bcl X|Apoptosis regulator Bcl-X|BCL2L|Bcl2-L-1|Bcl2 Like 1|Bcl-2-like protein 1|Bcl xL|BCL XL/S|Bcl xS|BclXs|B cell lymphoma 2 like|Bcl2l1|PPP1R52|DKFZp781P2092
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Synthesized peptide
- Purification:Affinity purified
- Cat. No.:77716-030
- Packaging:Vial
WB:1:500-1:1000, IHC:1:50-1:100, IF:1:100-1:200
Type:
Antigen: BCL2L1 (phospho–Ser62)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat