Anti-DNAJC15 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: DNAJC15
Clonality: Polyclonal
Clone:
Conjucation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77714-307
- 77714-308
- Antigen Synonyms:
- DJC15_HUMAN|Methylation controlled J protein|Cell growth inhibiting gene 22 protein|DnaJ homolog subfamily C member 15|Methylation-controlled J protein|DNAJ domain containing|DnaJ (Hsp40) homolog subfamily C member 15|DNAJC15|GIG22|DNAJD1|MCJ|HSD18|Cell growth-inhibiting gene 22 protein
- DJC15_HUMAN|Methylation controlled J protein|Cell growth inhibiting gene 22 protein|DnaJ homolog subfamily C member 15|Methylation-controlled J protein|DNAJ domain containing|DnaJ (Hsp40) homolog subfamily C member 15|DNAJC15|GIG22|DNAJD1|MCJ|HSD18|Cell growth-inhibiting gene 22 protein
- Gene ID:
- Q9Y5T4
- Q9Y5T4
- Immunogen:
- Recombinant Human DnaJ homolog subfamily C member 15 protein (59-150AA)
- Recombinant Human DnaJ homolog subfamily C member 15 protein (59-150AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice