Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Anti-NDUFAF1 Rabbit Polyclonal Antibody

Anti-NDUFAF1 Rabbit Polyclonal Antibody
 
Anti-NDUFAF1 Rabbit Polyclonal Antibody
Anti-NDUFAF1 Rabbit Polyclonal Antibody

 

 

ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: NDUFAF1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

 

 

  • Catalog No:
  • 77714-204
  • 77714-203
  • Antigen Symbol:
  • NDUFAF1
  • NDUFAF1
  • Antigen Synonyms:
  • NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 1|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1|Complex I intermediate associated protein 30 mitochondrial|Complex I intermediate-associated protein 30|NADH:ubiquinone oxidoreductase complex assembly factor 1|mitochondrial|CGI 65|CGI65|CIA30|Complex I intermediate associated protein 30|NDUFAF 1|CIA30_HUMAN|Ndufaf1
  • NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 1|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1|Complex I intermediate associated protein 30 mitochondrial|Complex I intermediate-associated protein 30|NADH:ubiquinone oxidoreductase complex assembly factor 1|mitochondrial|CGI 65|CGI65|CIA30|Complex I intermediate associated protein 30|NDUFAF 1|CIA30_HUMAN|Ndufaf1
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human|Mouse
  • Human|Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9Y375
  • Q9Y375
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Fusion protein of Human NDUFAF1
  • Fusion protein of Human NDUFAF1
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Product Family Options

 

  • Size
    Packaging
     
     
  • 50 µl
    Vial
       
       
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $230.89
    $230.89
    Each(1/Itm/Bx)
       $230.89
     

     : A48174-50UL
     
     
     
    Specifications
    Cat. No.
    77714-204
    Antigen Symbol
    NDUFAF1
    Antigen Synonyms
    NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 1|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1|Complex I intermediate associated protein 30 mitochondrial|Complex I intermediate-associated protein 30|NADH:ubiquinone oxidoreductase complex assembly factor 1|mitochondrial|CGI 65|CGI65|CIA30|Complex I intermediate associated protein 30|NDUFAF 1|CIA30_HUMAN|Ndufaf1
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human|Mouse
    Host
    Rabbit
    Gene ID
    Q9Y375
    Isotype
    IgG
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Immunogen
    Fusion protein of Human NDUFAF1
    Form
    Liquid
    Purification
    Antigen affinity purified
    Storage Buffer
    PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
    Storage Temperature
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
    Shipping Temperature
    Shipped on blue ice
  • 100 µl
    Vial
       
       
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $338.63
    $338.63
    Each(1/Itm/Bx)
       $338.63
     

     : A48174-100UL
     
     
     
    Specifications
    Cat. No.
    77714-203
    Antigen Symbol
    NDUFAF1
    Antigen Synonyms
    NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 1|NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1|Complex I intermediate associated protein 30 mitochondrial|Complex I intermediate-associated protein 30|NADH:ubiquinone oxidoreductase complex assembly factor 1|mitochondrial|CGI 65|CGI65|CIA30|Complex I intermediate associated protein 30|NDUFAF 1|CIA30_HUMAN|Ndufaf1
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human|Mouse
    Host
    Rabbit
    Gene ID
    Q9Y375
    Isotype
    IgG
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Immunogen
    Fusion protein of Human NDUFAF1
    Form
    Liquid
    Purification
    Antigen affinity purified
    Storage Buffer
    PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
    Storage Temperature
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
    Shipping Temperature
    Shipped on blue ice