Anti-AGA Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: AGA
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77713-635
- 77713-634
- Antigen Symbol:
- AGA
- AGA
- Antigen Synonyms:
- ASPG_HUMAN|Aspartylglucosylamine deaspartylase|N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase|N4 (N acetyl beta glucosaminyl) L asparagine amidase|N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase|AGU|Aspartylglucosaminidase|Glycosylasparaginase|Aga|GA|Glycosylasparaginase beta chain|ASRG|Aspartylglycosaminuria
- ASPG_HUMAN|Aspartylglucosylamine deaspartylase|N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase|N4 (N acetyl beta glucosaminyl) L asparagine amidase|N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase|AGU|Aspartylglucosaminidase|Glycosylasparaginase|Aga|GA|Glycosylasparaginase beta chain|ASRG|Aspartylglycosaminuria
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20933
- P20933
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human AGA
- Fusion protein of Human AGA
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice