Anti-STK17A Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: STK17A
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77713-122
- 77713-121
- Antigen Symbol:
- STK17A
- STK17A
- Antigen Synonyms:
- Serine/threonine-protein kinase 17A|Serine/threonine kinase 17a|STK17A|EC 2.7.11.1|ST17A_HUMAN|DRAK1|death-associated protein kinase-related 1|STK17 A|Serine/threonine protein kinase 17a|Serine / threonine kinase 17a death associated protein kinase related 1|DAP kinase-related apoptosis-inducing protein kinase 1
- Serine/threonine-protein kinase 17A|Serine/threonine kinase 17a|STK17A|EC 2.7.11.1|ST17A_HUMAN|DRAK1|death-associated protein kinase-related 1|STK17 A|Serine/threonine protein kinase 17a|Serine / threonine kinase 17a death associated protein kinase related 1|DAP kinase-related apoptosis-inducing protein kinase 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UEE5
- Q9UEE5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human STK17A
- Synthetic peptide of Human STK17A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice