Anti-SIRT4 Rabbit Polyclonal Antibody
New Product
Type:
Antigen: SIRT4
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77713-037
- 77713-038
- Antigen Symbol:
- SIRT4
- SIRT4
- Antigen Synonyms:
- MGC130046|Sirt4|Regulatory protein SIR2 homolog 4|SIR2L 4|NAD-dependent protein lipoamidase sirtuin-4, mitochondrial|NAD dependent protein deacetylase sirtuin 4|SIR2-like protein 4|Sirtuin (silent mating type information regulation 2 homolog) 4|Sir2 like 4|Sirtuin (silent mating type information regulation 2 homolog) 4 (S. cerevisiae)|Silent mating type information regulation 2 homolog|SIR2L4|SIR 2 like protein 4|Sir 2 like 4|NAD dependent deacetylase sirtuin 4|Sirtuin-4|Sirtuin4|MGC130047|SIR2 like protein 4|SIRT 4|NAD-dependent protein deacetylase sirtuin-4|MGC57437|Sirtuin 4|Sirtuin type 4|SIR4_HUMAN|NAD-dependent ADP-ribosyltransferase sirtuin-4
- MGC130046|Sirt4|Regulatory protein SIR2 homolog 4|SIR2L 4|NAD-dependent protein lipoamidase sirtuin-4, mitochondrial|NAD dependent protein deacetylase sirtuin 4|SIR2-like protein 4|Sirtuin (silent mating type information regulation 2 homolog) 4|Sir2 like 4|Sirtuin (silent mating type information regulation 2 homolog) 4 (S. cerevisiae)|Silent mating type information regulation 2 homolog|SIR2L4|SIR 2 like protein 4|Sir 2 like 4|NAD dependent deacetylase sirtuin 4|Sirtuin-4|Sirtuin4|MGC130047|SIR2 like protein 4|SIRT 4|NAD-dependent protein deacetylase sirtuin-4|MGC57437|Sirtuin 4|Sirtuin type 4|SIR4_HUMAN|NAD-dependent ADP-ribosyltransferase sirtuin-4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6E7
- Q9Y6E7
- Isotype:
- IgG
- IgG
- Immunogen:
- Fusion protein of Human SIRT4
- Fusion protein of Human SIRT4
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice