Anti-USP15 Rabbit Polyclonal Antibody
New Product
ELISA:1:2000-1:5000, IHC:1:25-1:100
Type:
Antigen: USP15
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77713-013
- 77713-012
- Antigen Symbol:
- USP15
- USP15
- Antigen Synonyms:
- UBP15_HUMAN|Ubiquitin Specific Protease 15|Deubiquitinating enzyme|UBP 15|Ubiquitin specific processing protease 15|Ubiquitin Carboxy terminal Hydrolase 15|Ubiquitin thioesterase 15|Usp15|Ubiquitin thiolesterase 15|MGC131982|UNPH 4|UBP15|Ubiquitin carboxyl-terminal hydrolase 15|Unph2|MGC149838|Ubiquitin specific peptidase 15|Ubiquitin-specific-processing protease 15|Unph4|KIAA0529|USP 15|Unph-2|EC 3 1 2 15|Unph 2|Deubiquitinating enzyme 15|MGC74854
- UBP15_HUMAN|Ubiquitin Specific Protease 15|Deubiquitinating enzyme|UBP 15|Ubiquitin specific processing protease 15|Ubiquitin Carboxy terminal Hydrolase 15|Ubiquitin thioesterase 15|Usp15|Ubiquitin thiolesterase 15|MGC131982|UNPH 4|UBP15|Ubiquitin carboxyl-terminal hydrolase 15|Unph2|MGC149838|Ubiquitin specific peptidase 15|Ubiquitin-specific-processing protease 15|Unph4|KIAA0529|USP 15|Unph-2|EC 3 1 2 15|Unph 2|Deubiquitinating enzyme 15|MGC74854
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4E8
- Q9Y4E8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human USP15
- Fusion protein of Human USP15
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together