Major Cations Mix Standard: Al, Ca, Fe, K, Mg, Na @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-851
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:1000 µg/mL
- Matrix:5% HNO₃
- Description:Major Cations Mix Standard: Al, Ca, Fe, K, Mg, Na @ 1000 µg/mL in 5% HNO3
- Size:100 ml
- Cat. No.:77697-851