Order Entry
Puerto Rico
ContactUsLinkComponent
 
Scandium Standard: Sc @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-911
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Scandium Standard: Sc @ 10000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-911
Supplier:  LGC Standards
Supplier Number:  VHG-TSCN-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10000 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Scandium Standard: Sc @ 10000 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-911

Specifications

About this item