Order Entry
Puerto Rico
ContactUsLinkComponent
 
Metalloids/Hydride Elements Standard: As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/mL in 20% HCl, tr. HF, VHG, LGC Standards
Catalog # 77697-858
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Metalloids/Hydride Elements Standard: As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/mL in 20% HCl, tr. HF, VHG, LGC Standards
Catalog # 77697-858
Supplier:  LGC Standards
Supplier Number:  VHG-SM50B-100
New Product
CAS Number:  

Specifications

  • Concentration:
    100 µg/mL
  • Matrix:
    20% HCl
  • Description:
    Metalloids/Hydride Elements Standard: As, Bi, Ga, Ge, In, Pb, Sb, Se, Sn, Te, Tl @ 100 µg/mL in 20% HCl, tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77697-858

Specifications

About this item