Order Entry
Puerto Rico
ContactUsLinkComponent
Major Environmental Elements: Na, Ca, K, Mg @ 1000 µg/mL in 2% HNO3, VHG, LGC Standards
Major Environmental Elements: Na, Ca, K, Mg @ 1000 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-857
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Major Environmental Elements: Na, Ca, K, Mg @ 1000 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77697-857
Supplier:  LGC Standards
Supplier Number:  VHG-SM4-1000-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    Major Environmental Elements: Na, Ca, K, Mg @ 1000 µg/mL in 2% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-857

Specifications

About this item