Refractory Elements Standard: Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 µg/mL in 5% HCl, tr. HF, VHG, LGC Standards
Catalog # 77697-854
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:5% HCl/tr HF
- Description:Refractory Elements Standard: Al, B, Cr, Hf, Mo, Nb, Si, Ta, Ti, V, W, Zr @ 100 µg/mL in 5% HCl, tr. HF
- Size:100 ml
- Cat. No.:77697-854