Order Entry
Puerto Rico
ContactUsLinkComponent
Manganese - Mn @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Manganese - Mn @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77697-714
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Manganese - Mn @ 5000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77697-714
Supplier:  LGC Standards
Supplier Number:  VHG-OMN-5000-A-50G
New Product
CAS Number:  

Specifications

  • Concentration:
    5000 µg/mL
  • Matrix:
    oil
  • Description:
    Manganese - Mn @ 5000 µg/g in Hydrocarbon Oil
  • Size:
    50 ml
  • Cat. No.:
    77697-714

Specifications

About this item