Order Entry
Puerto Rico
ContactUsLinkComponent
MISA Standard 3: Sb, Sn, Te @ 100 µg/mL in 20% HCl, VHG, LGC Standards
MISA Standard 3: Sb, Sn, Te @ 100 µg/mL in 20% HCl, VHG, LGC Standards
Catalog # 77697-691
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
MISA Standard 3: Sb, Sn, Te @ 100 µg/mL in 20% HCl, VHG, LGC Standards
Catalog # 77697-691
Supplier:  LGC Standards
Supplier Number:  VHG-MISA3-100
New Product
CAS Number:  

Specifications

  • Concentration:
    100 µg/mL
  • Matrix:
    20% HCl
  • Description:
    MISA Standard 3: Sb, Sn, Te @ 100 µg/mL in 20% HCl
  • Size:
    100 ml
  • Cat. No.:
    77697-691

Specifications

About this item