Order Entry
Puerto Rico
ContactUsLinkComponent
Lead Standard: Pb @ 1.0 g/gal in Isooctane, VHG, LGC Standards
Lead Standard: Pb @ 1.0 g/gal in Isooctane, VHG, LGC Standards
Catalog # 77697-742
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Lead Standard: Pb @ 1.0 g/gal in Isooctane, VHG, LGC Standards
Catalog # 77697-742
Supplier:  LGC Standards
Supplier Number:  VHG-PBISO-1-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1.0 g/gal
  • Description:
    Lead Standard: Pb @ 1.0 g/gal in Isooctane
  • Size:
    100 ml
  • Cat. No.:
    77697-742

Specifications

About this item