Order Entry
Puerto Rico
ContactUsLinkComponent
ICP-MS Setup Solution: Be, Ce, Fe, In, Li, Mg, Pb, U @ 1 µg/L in 1% HNO3, VHG, LGC Standards
ICP-MS Setup Solution: Be, Ce, Fe, In, Li, Mg, Pb, U @ 1 µg/L in 1% HNO3, VHG, LGC Standards
  77697-665
New Product
 :  
undefined
ICP-MS Setup Solution: Be, Ce, Fe, In, Li, Mg, Pb, U @ 1 µg/L in 1% HNO3, VHG, LGC Standards
  77697-665
 :  VHG-LPENXSUSDIL-500
New Product
 :  

 

  • Concentration:
    1 µg/L
  • Matrix:
    1% HNO₃
  • Description:
    ICP-MS Setup Solution: Be, Ce, Fe, In, Li, Mg, Pb, U @ 1 µg/L in 1% HNO3
  • Size:
    500 ml
  • Cat. No.:
    77697-665