Order Entry
Puerto Rico
ContactUsLinkComponent
ICP-MS Internal Standard: Bi @ 100 ug/mL in 2% HNO3, VHG, LGC Standards
ICP-MS Internal Standard: Bi @ 100 ug/mL in 2% HNO3, VHG, LGC Standards
  77697-635
New Product
 :  
undefined
ICP-MS Internal Standard: Bi @ 100 ug/mL in 2% HNO3, VHG, LGC Standards
  77697-635
 :  VHG-LISBI100-100
New Product
 :  

 

  • Concentration:
    100 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    ICP-MS Internal Standard: Bi @ 100 ug/mL in 2% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-635