Silica Standard: SiO2 @ 100 µg/mL in H2O, VHG, LGC Standards
Catalog # 77697-593
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:H₂O
- Description:Silica Standard: SiO2 @ 100 µg/mL in H2O
- Size:500 ml
- Cat. No.:77697-593