Order Entry
Puerto Rico
ContactUsLinkComponent
Mercury ICV Standard (Second Source): Hg @ 10 µg/mL in 5% HNO3, VHG, LGC Standards
Mercury ICV Standard (Second Source): Hg @ 10 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-592
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Mercury ICV Standard (Second Source): Hg @ 10 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-592
Supplier:  LGC Standards
Supplier Number:  VHG-ISHG-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Mercury ICV Standard (Second Source): Hg @ 10 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-592

Specifications

About this item