ICH/USP Parenteral Combined-2 Target Elements: Au @ 100; Ir, Os, Pd, Pt, Rh, Ru @ 10 µg/mL in 15% HCl, VHG, LGC Standards
Catalog # 77697-541
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:15% HCl
- Description:ICH/USP Parenteral Combined-2 Target Elements: Au @ 100; Ir, Os, Pd, Pt, Rh, Ru @ 10 µg/mL in 15% HCl
- Size:100 ml
- Cat. No.:77697-541